Learn More
enQuireBio™ Recombinant Human NTHL1 Protein
A cDNA sequence encoding the NTHL1 was constructed and used to recombinantly synthesize the protein.
Marke:  enQuireBio™ QP12893-2ug
 Lernen Sie weitere Varianten dieses
Artikelnummer. 15991869
  • 141.00 CHF / 2 Mikrogramm
Verfügbar ab: 09-05-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Spezifikation
4913
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MCSPQESGMT ALSARMLTRS RSLGPGAGPR GCREEPGPLR RREAAAEARK SHSPVKRPRK AQRLRVAYEG SDSEKGEGAE PLKVPVWEPQ DWQQQLVNIR AMRNKKDAPV DHLGTEHCYD SSAPPKVRRY QVLLSLMLSS QTKDQVTAGA MQRLRARGLT VDSILQTDDA TLGKLIYPVG FWRSKVKYIK QTSAILQQHY GGDIPASVAE LVALPGVGPK MAHLAMAVAW GTVSGIAVDT HVHRIANRLR WTKKATKSPE ETRAALEEWL PRELWHEING LLVGFGQQTC LPVHPRCHAC LNQALCPAAQ GL
Greater than 80.0% as determined by SDS-PAGE.
NTHL1 Protein
Research Use Only
NTHL1
Human
His
NTHL1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 40% glycerol, 0.15M NaCl and 1mM DTT.
Dokumentation
One moment while we fetch your results.
Name des Produkts
enQuireBio™ Recombinant Human NTHL1 Protein > 2μg