missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NTHL1 Protein
A cDNA sequence encoding the NTHL1 was constructed and used to recombinantly synthesize the protein.
141.00 CHF - 8085.00 CHF
Spezifikation
Gen-ID (Entrez) | 4913 |
---|---|
Name | NTHL1 Protein |
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gensymbol | NTHL1 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | ||||||
15991869
|
enQuireBio™
QP12893-2ug |
2 μg |
141.00 CHF
2 Mikrogramm |
Verfügbar ab: 09-05-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
||||||
15971869
|
enQuireBio™
QP12893-10ug |
10 μg |
219.00 CHF
10 Mikrogramm |
Verfügbar ab: 09-05-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
||||||
15981869
|
enQuireBio™
QP12893-1mg |
1 mg |
8085.00 CHF
1 Milligramm |
Verfügbar ab: 09-05-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
||||||
Spécification
4913 | |
Research Use Only | |
NTHL1 | |
Human | |
His | |
NTHL1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 40% glycerol, 0.15M NaCl and 1mM DTT. |
NTHL1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MCSPQESGMT ALSARMLTRS RSLGPGAGPR GCREEPGPLR RREAAAEARK SHSPVKRPRK AQRLRVAYEG SDSEKGEGAE PLKVPVWEPQ DWQQQLVNIR AMRNKKDAPV DHLGTEHCYD SSAPPKVRRY QVLLSLMLSS QTKDQVTAGA MQRLRARGLT VDSILQTDDA TLGKLIYPVG FWRSKVKYIK QTSAILQQHY GGDIPASVAE LVALPGVGPK MAHLAMAVAW GTVSGIAVDT HVHRIANRLR WTKKATKSPE ETRAALEEWL PRELWHEING LLVGFGQQTC LPVHPRCHAC LNQALCPAAQ GL | |
Greater than 80.0% as determined by SDS-PAGE. |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
enQuireBio™ Recombinant Human NTHL1 Protein