Learn More
enQuireBio™ Recombinant Human NTHL1 Protein
A cDNA sequence encoding the NTHL1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12893-1mg
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
4913 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MCSPQESGMT ALSARMLTRS RSLGPGAGPR GCREEPGPLR RREAAAEARK SHSPVKRPRK AQRLRVAYEG SDSEKGEGAE PLKVPVWEPQ DWQQQLVNIR AMRNKKDAPV DHLGTEHCYD SSAPPKVRRY QVLLSLMLSS QTKDQVTAGA MQRLRARGLT VDSILQTDDA TLGKLIYPVG FWRSKVKYIK QTSAILQQHY GGDIPASVAE LVALPGVGPK MAHLAMAVAW GTVSGIAVDT HVHRIANRLR WTKKATKSPE ETRAALEEWL PRELWHEING LLVGFGQQTC LPVHPRCHAC LNQALCPAAQ GL | |
Greater than 80.0% as determined by SDS-PAGE. |
NTHL1 Protein | |
Research Use Only | |
NTHL1 | |
Human | |
His | |
NTHL1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 40% glycerol, 0.15M NaCl and 1mM DTT. |
Berichtigung von Produktinhalten
Name des Produkts
enQuireBio™ Recombinant Human NTHL1 Protein > 1mg
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur