Learn More
alpha-Methylacyl-CoA Racemase/AMACR Antibody (AMACR/4572R), Alexa Fluor™ 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Marke: Novus Biologicals NBP3-08468AF350
Beschreibung
alpha-Methylacyl-CoA Racemase/AMACR Monoclonal antibody specifically detects alpha-Methylacyl-CoA Racemase/AMACR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Spezifikation
alpha-Methylacyl-CoA Racemase/AMACR | |
Monoclonal | |
Alexa Fluor 350 | |
2-methylacyl-CoA racemase, alpha-methylacyl-CoA racemase, CBAS4, EC 5.1.99.4, RACE, RM | |
Recombinant full-length human alpha-Methylacyl-CoA Racemase/AMACR protein (Uniprot: Q9UHK6) | |
100 μg | |
Lipid and Metabolism, Prostate Cancer, Signal Transduction | |
23600 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
AMACR/4572R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Produktvorschläge
Kunden, die diesen Artikel ansahen, interessierten sich auch für
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.