Learn More
Novus Biologicals™ Recombinant Human NNT GST (N-Term) Protein
Recombinant Protein
Marke: Novus Biologicals™ H00023530-P01-25ug
Beschreibung
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Spezifikation
Western Blot, ELISA, Protein Array, Immunoaffinity Purification | |
23530 | |
25 μg | |
MGC126502, mitochondrial, nicotinamide nucleotide transhydrogenaseMGC126503, Pyridine nucleotide transhydrogenase | |
>80% by SDS-PAGE and Coomassie blue staining |
50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. | |
MolecularWeight-theroretical: 48.51 kDa | |
Store at -80°C. Avoid freeze-thaw cycles. | |
NNT | |
NNT |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.