Learn More
Thermo Scientific™ Nalgene™Polycarbonate Biotainer™Bottles and Carboys, 5L Artikelnummer.: 15656638
Thermo Scientific™ Nalgene™Polycarbonate Biotainer™Bottles and Carboys, 5L
Artikelnummer. 15656638
Artikelnummer. 15656638
Marke: Thermo Scientific™ 38340542x
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.
Alternative Produkte anzeigen
Specifically designed for pharmaceutical and biological manufacturing to reduce the risk of carry-over contamination. Durable, translucent PC construction ensures the integrity of your sample, even frozen to -100°C. 
Provided sterile and ready-to-use; including certified clean products, which meet USP <788> standards and endotoxins below 0.5 EU/mL. 
Specifically designed for pharmaceutical and biological manufacturing, Thermo Scientific™
Nalgene™ Polycarbonate Biotainer™ Bottles and Carboys reduce the risk of carry-over
contamination. Durable, translucent PC construction ensures the integrity of your sample, even
frozen to -100°C.
  • Provided sterile and ready-to-use; including certified clean products, which meet USP<788> standards and endotoxins below 0.5 EU/mL
  • Certified clean products include triple layer packaging, with two heat-sealed layers to facilitate ease of  entry into cleanrooms
  • Certified clean products include triple layer packaging, with two heat-sealed layers to facilitate ease of entry into cleanrooms
  • Sterile, ready-to-use products with a Sterility Assurance Level of 10-6 inside the bottle/closure and two heat-sealed sterile barriers to protect product prior to use
  • All heat-sealed bags include quick-tears on the top and bottom and placed in carton liner twisted and tape/tie sealed to prevent gross contamination on the inner packaging
  • Certified to meet USP <788> low particulate requirements to limit potential particulate contamination
  • Certified low endotoxin (0.50 EU/mL) according to USP<85> to reduce pyrogens
  • Products can be supplied with forced extraction studies*, validation binders* and certificates
  • Inspection procedures throughout manufacturing include leak testing, dimensional testing, visual inspection for particulates and release testing to ensure the product meets all specifications
  • Clean roomproduced in ISO Class 7 to limit particulates during molding 
  • All container/closure systems are certified leakproof** to protect valuable contents
  • Robust, PC can be used from -100˚C to 100˚C, allowing for both heated mixing and frozen storage or shipment. Must be tested in actual use conditions as results may vary depending upon application.
  • Customer support includes our highly experienced commercial specialist teams, regulatory team, quality team and regional technical support
TRUSTED_SUSTAINABILITY
Zertifikate
Eine Chargennummer ist erforderlich, um Ergebnisse für Zertifikate anzuzeigen. Um Ihre Chargennummer zu früheren Bestellungen zu finden, können Sie unsere Auftragsverfolgung nutzen.
5 Ergebnisse gefunden
Chargennummer Art des Zertifikats Datum Catalog Number
Chargennummer1306166 Art des ZertifikatsKonformitätsbescheinigung Datum12/12/2024 Catalog Number383405-42,383405-42X
Chargennummer1304547 Art des ZertifikatsKonformitätsbescheinigung Datum11/12/2024 Catalog Number383405-42,383405-42X
Chargennummer1381606 Art des ZertifikatsVerarbeitungszertifikat Datum04/12/2024 Catalog Number383405-42,383405-42X
Chargennummer1263808 Art des ZertifikatsVerarbeitungszertifikat Datum04/12/2024 Catalog Number383405-42,383405-42X
Chargennummer1396653 Art des ZertifikatsVerarbeitungszertifikat Datum04/12/2024 Catalog Number383405-42,383405-42X
Berichtigung von Produktinhalten
Name des Produkts
Thermo Scientific™ Nalgene™Polycarbonate Biotainer™Bottles and Carboys, 5L >