missing translation for 'onlineSavingsMsg'
Learn More
Maraviroc, Tocris Bioscience™ Artikelnummer.: 18094942
Verfügbar ab: 27-05-2025
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Maraviroc, Tocris Bioscience™

Product Code. 18094942
50 mg
missing translation for 'orderingAttributeHoverText'
Menge:
10 mg
50 mg
This item is not returnable. View return policy

Artikelnummer. 18094942

Marke: Tocris Bioscience 3756/50

Verfügbar ab: 27-05-2025
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
This item is not returnable. View return policy

Selective CCR5 antagonist

Selective CCR5 antagonist; displays potent anti-HIV-1 activity. Prevents the interaction of HIV-1 gp120 and CCR5 (IC50 = 6.4 nM), inhibiting HIV-1 entry. Exhibits antinociceptive effects in a rat model of neuropathic pain. Also inhibits CCL3 (MIP-1α) binding to CCR5. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 376348-65-1
Summenformel C29H41F2N5O
Molekulargewicht (g/mol) 513.678
InChI-Schlüssel GSNHKUDZZFZSJB-HLMSNRGBSA-N
Synonym maraviroc, selzentry, celsentri, maraviroc usan, celsentri tm, selzentry tm, 3h maraviroc, unii-md6p741w8a, pro 140 & maraviroc, mvcMehr anzeigenWeniger anzeigen
PubChem CID 3002977
IUPAC-Name 4,4-difluoro-N-[(1S)-3-[(1S,5R)-3-(3-methyl-5-propan-2-yl-1,2,4-triazol-4-yl)-8-azabicyclo[3.2.1]octan-8-yl]-1-phenylpropyl]cyclohexane-1-carboxamideMehr anzeigenWeniger anzeigen
SMILES CC1=NN=C(N1C2CC3CCC(C2)N3CCC(C4=CC=CC=C4)NC(=O)C5CCC(CC5)(F)F)C(C)C

Specifications

Chemischer Name oder Material Maraviroc
Menge 50 mg
Formelmasse 513.67
Reinheit (%) >99%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Maraviroc, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.