Learn More
Heidolph™ Titramax 1000
Artikelnummer. 17373470 Alle ansehen Heidolph Instruments Produkte
CH-Stecker, 1 Stück
Click to view available options
Steckertyp:
CH-Stecker
Packungsgröße:
1 Stück
Artikelnummer. 17373470
Marke: Heidolph™ 5441220002
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Titramax 1000 is the largest model of the series and contains all the features of the Titramax 100.
  • The largest model of the Titramax series contains all the features of the Titramax 100: a shaking orbit of 1.5 mm, speeds of up to 1350 rpm, and a timer.
  • With a storage capacity of up to six multi-well plates, a higher number of samples can be processed. The option to integrate the Titramax 1000 into the modular incubation system also allows temperature-controlled processes to be carried out without problems.
  • With this model, easy loading and removal of the plates can be carried out comfortably due to the recess and the spacings in the rubber mat.
TRUSTED_SUSTAINABILITY
Spezifikation
Spannung 230 V
Steckertyp CH-Stecker
Berichtigung von Produktinhalten
Name des Produkts
Heidolph™ Titramax 1000 >