Learn More
RAI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech 17084-1-AP-150UL
 Lernen Sie weitere Varianten dieses
Artikelnummer. 16806924
  • 385.92 CHF / 150 Mikroliter
Lager vorrätig
Entdecken Sie weitere Sonderangebote
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
This gene is located within the Smith-Magenis syndrome region on chromosome 17. It is highly similar to its mouse counterpart and is expressed at high levels mainly in neuronal tissues. The protein encoded by this gene includes a polymorphic polyglutamine tract in the N-terminal domain. Expression of the mouse counterpart in neurons is induced by retinoic acid. This gene is associated with both the severity of the phenotype and the response to medication in schizophrenic patients.
Spezifikation
Spezifikation
RAI1
Polyclonal
Unconjugated
RAI1
KIAA1820, RAI1, retinoic acid induced 1, SMCR, SMS
Rabbit
Antigen Affinity Chromatography
RUO
10743, 19377, 303188
-20°C
Liquid
Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q61818, Q7Z5J4
Rai1
RAI1 Fusion Protein Ag9046
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
One moment while we fetch your results.
Zertifikate
Sonderangebote
Sonderangebote
Berichtigung von Produktinhalten
Name des Produkts
RAI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated