Learn More
Novus Biologicals™ IKK gamma Inhibitor Peptide Set
Applications: In vitro assay, In vivo assay, Functional (Inhibition), Block/Neutralize
Marke: Novus Biologicals™ NBP2-26504-5MG
Beschreibung

Spezifikation
Human, Mouse, Rat | |
Functional (Inhibition), In vitro assay, In vivo assay | |
5 mg | |
IKK gamma Inhibitor |
1.IKK-gamma NEMO Binding Domain (NBD) Inhibitor Peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKKTALDWSWLQTE (IKK-gamma/NEMO binding sequence is underlined). Molecular weight: 3692 2.Antennapedia Control peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKK. Molecular weight: 2361 |
|
Store at -20°C. Avoid freeze-thaw cycles. | |
3692 |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only