Learn More
BEX1/2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12390-1-AP-20UL
Beschreibung
This antibody detects both BEX1 and BEX2.
BEX1/BEX2 belongs to the brain expressed X-linked gene family. The encoded protein interacts with the transcription factor LIM domain only 2 in a DNA-binding complex that recognizes the E-box element and promotes transcription. BEX1/BEX2 has been found to be a tumor suppressor that is silenced in human glioma. In breast cancer cells, this gene product modulates apoptosis in response to estrogen and tamoxifen, and enhances the anti-proliferative effect of tamoxifen. Multiple transcript variants encoding different isoforms have been found for this gene.Spezifikation
BEX1/2 | |
Polyclonal | |
Unconjugated | |
BEX1 | |
BEX1, BEX1/2, BEX2, brain expressed X linked 2, hBex2, Protein BEX2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12069, 19716, 363498, 501625, 55859, 84707 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3MKQ1, Q3MKQ2, Q9BXY8, Q9HBH7, Q9R224, Q9WTZ8 | |
Bex1, Bex2 | |
BEX1/2 Fusion Protein Ag3066 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.