missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCA1 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NB400-105JF646
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
ABCA1 Polyclonal antibody specifically detects ABCA1 in Human, Mouse, Rat, Porcine, Canine, Chicken, Chinese Hamster, Equine, Hamster, Mustelid, Primate, Rabbit samples. It is validated for Western Blot, Simple Western, ChIP assay, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Gel Super Shift Assays, Gel Supershift Assay, ChIP assay (ChIP), Gene Knock-Out, KnockDown, Neutralization, PCR, Dual RNAscope ISH-IHC
Spezifikation
ABCA1 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Porcine, Canine | |
Purified |
Western Blot, Simple Western, ChIP Assay, Flow Cytometry, ELISA | |
Janelia Fluor 646 | |
ABC-1, ABC1ATP-binding cassette transporter A1, ATP-binding cassette 1, ATP-binding cassette transporter 1, ATP-binding cassette, sub-family A (ABC1), member 1, CERP, Cholesterol efflux regulatory protein, EC 2.7.7.8, EC 2.8.1.8, EC 3.6.3, EC 3.6.3.41, FLJ14958, HDLDT1, membrane-bound, MGC164864, MGC165011, TGDATP-binding cassette sub-family A member 1 | |
Partial peptide sequence (within residues 1100-1300) of human ABCA1 Antibody [UniProt# O95477]. Actual immunogen sequence is proprietary information. | |
0.1 mL | |
ABC Transporters, Astrocyte Markers, Cancer, Cardiovascular Biology, Cellular Markers, Cholesterol Metabolism, Diabetes Research, Glia Markers, Golgi Apparatus Markers, Lipid and Metabolism, Neuroscience, Signal Transduction | |
19 | |
Store at 4°C in the dark. | |
IgG |
Produktvorschläge
Kunden, die diesen Artikel ansahen, interessierten sich auch für
Viewing 1-4 of
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
ABCA1 Antibody, Janelia Fluor™ 646, Novus Biologicals™ > 0.1 mL; Janelia Fluor 646
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur