CAS RN 10034-85-2
CAS RN 10034-85-2
Iodwasserstoffsäure, zur Analyse, 57 Gew.% Lösung in Wasser, dest., stabilisiert, Thermo Scientific Chemicals
CAS: 10034-85-2 Summenformel: HI Molekulargewicht (g/mol): 127.91 MDL-Nummer: MFCD00011347 InChI-Schlüssel: XMBWDFGMSWQBCA-UHFFFAOYSA-N IUPAC-Name: Wasserstoffiodid SMILES: I
Iodwasserstoffsäure, zur Analyse, 57 Gew.% Lösung in Wasser, dest., ohne Stabilisator, Thermo Scientific Chemicals
CAS: 10034-85-2 Summenformel: HI Molekulargewicht (g/mol): 127.91 MDL-Nummer: MFCD00011347 InChI-Schlüssel: XMBWDFGMSWQBCA-UHFFFAOYSA-N IUPAC-Name: Wasserstoffiodid SMILES: I
Hydriodsäure, ACS, 55 bis 58 %, Thermo Scientific Chemicals
CAS: 10034-85-2 Summenformel: HI Molekulargewicht (g/mol): 127.91 MDL-Nummer: MFCD00011347 InChI-Schlüssel: XMBWDFGMSWQBCA-UHFFFAOYSA-N IUPAC-Name: hydrogen iodide SMILES: I
Iodwasserstoffsäure, ACS Reagenz, 55 % in Wasser, ohne Stabilisator, Thermo Scientific Chemicals
CAS: 10034-85-2 Summenformel: HI Molekulargewicht (g/mol): 127.91 MDL-Nummer: MFCD00011347 InChI-Schlüssel: XMBWDFGMSWQBCA-UHFFFAOYSA-N IUPAC-Name: hydrogen iodide SMILES: I
Hydriodsäure, ACS, 47 %, stab. mit 1,5 % Hypophosphorsäure, Thermo Scientific Chemicals
CAS: 10034-85-2 Summenformel: HI
Hydriodic Acid, 55 to 58%, ACS Reagent, Honeywell™ Fluka™
CAS: 10034-85-2 Summenformel: HI Molekulargewicht (g/mol): 127.91 MDL-Nummer: MFCD00011347 InChI-Schlüssel: XMBWDFGMSWQBCA-UHFFFAOYSA-N Synonym: hydriodic acid,hydrogen iodide,hydroiodic acid,hydriotic acid,iodium,yodo,jodwasserstoff,iodine-125,unii-694c0eft9q,iodo group PubChem CID: 24841 ChEBI: CHEBI:43451 IUPAC-Name: hydrogen iodide SMILES: I
Hydriodic Acid, stabilized, Honeywell
CAS: 10034-85-2 Summenformel: HI Molekulargewicht (g/mol): 127.91 MDL-Nummer: MFCD00011347 InChI-Schlüssel: XMBWDFGMSWQBCA-UHFFFAOYSA-N Synonym: hydriodic acid,hydrogen iodide,hydroiodic acid,hydriotic acid,iodium,yodo,jodwasserstoff,iodine-125,unii-694c0eft9q,iodo group PubChem CID: 24841 ChEBI: CHEBI:43451 IUPAC-Name: hydrogen iodide SMILES: I
Hydriodic Acid, no stabilizer, Honeywell
CAS: 10034-85-2 Summenformel: HI Molekulargewicht (g/mol): 127.91 MDL-Nummer: MFCD00011347 InChI-Schlüssel: XMBWDFGMSWQBCA-UHFFFAOYSA-N Synonym: hydriodic acid,hydrogen iodide,hydroiodic acid,hydriotic acid,iodium,yodo,jodwasserstoff,iodine-125,unii-694c0eft9q,iodo group PubChem CID: 24841 ChEBI: CHEBI:43451 IUPAC-Name: hydrogen iodide SMILES: I
Hydriodic Acid, Puriss p.a., ≥67% (T), Honeywell™
CAS: 10034-85-2 Summenformel: HI Molekulargewicht (g/mol): 127.91 MDL-Nummer: MFCD00011347 InChI-Schlüssel: XMBWDFGMSWQBCA-UHFFFAOYSA-N Synonym: hydriodic acid,hydrogen iodide,hydroiodic acid,hydriotic acid,iodium,yodo,jodwasserstoff,iodine-125,unii-694c0eft9q,iodo group PubChem CID: 24841 ChEBI: CHEBI:43451 IUPAC-Name: hydrogen iodide SMILES: I