Learn More
Cytiva Whatman™ 3MM Chr Chromatography Paper
Manufactured entirely from high-quality cotton linters with no additives and tested specifically for chromatographic techniques. Whatman™ Grade 3MM Chr Chromatography Paper ensures the wicking capability and uniformity of capillary action that are important in chemical separations.
32.40CHF - 690.00CHF
Specifications
Description 0.34 mm Whatman medium thickness 3 MM Chr paper
Flow Rate 130 mm/30 min.
Film Thickness 0.34 mm
Finish Smooth
For Use With (Application) For Electrophoresis, General Chromatography, and Blotting AppliCatIons
View More Specs
Products 22
Product Code Brand Length (Metric) Width (Metric) Format Price Quantity & Availability  
Product Code Brand Length (Metric) Width (Metric) Format Price Quantity & Availability  
11330744
View Documents Promotion Details
Cytiva
3030-917
57 cm 46 cm Sheet
544.00CHF
Pack of 100
Only null left
In Stock
Edge
Add to basket
11895375
View Documents Promotion Details
Cytiva
3030-614
100 m 2 cm Roll
54.40CHF
Each
Only null left
In Stock
Add to basket
11348924
View Documents Promotion Details
Cytiva
3030-153
17.5 cm 15 cm Sheet
62.15CHF
Pack of 100
Only null left
In Stock
Add to basket
11975194
View Documents
Cytiva
3030-221
34 cm 18 cm Sheet
141.00CHF
Pack of 100
Only null left
In Stock
Add to basket
11919197
View Documents Promotion Details
Cytiva
3030-392
45 cm 35 cm Sheet
230.00CHF
Pack of 100
Only null left
Estimated Shipment: 17-06-2025
Add to basket
11358934
View Documents Promotion Details
Cytiva
3030-662
100 m 7.5 cm Roll
164.00CHF
Each
Only null left
In Stock
Add to basket
10441412
View Documents Promotion Details
Cytiva
3030-672
100 m 10 cm Roll
269.00CHF
Each
Only null left
In Stock
Add to basket
11338934
View Documents
Cytiva
3030-675
100 m 12.5 cm Roll
259.00CHF
Each
Only null left
Estimated Shipment: 23-10-2025
Add to basket
11380734
View Documents Promotion Details
Cytiva
3030-681
100 m 15 cm Roll
301.00CHF
Each
Only null left
In Stock
Add to basket
11390734
View Documents
Cytiva
3030-690
100 m 19 cm Roll
375.00CHF
Each
Only null left
In Stock
Add to basket
11300744
View Documents
Cytiva
3030-700
100 m 23 cm Roll
442.00CHF
Each
Only null left
In Stock
Add to basket
11310744
View Documents
Cytiva
3030-704
100 m 27 cm Roll
489.00CHF
Each
Only null left
Estimated Shipment: 23-10-2025
Add to basket
11320744
View Documents Promotion Details
Cytiva
3030-861
20 cm 20 cm Sheet
99.20CHF
Pack of 100
Only null left
In Stock
Edge
Add to basket
10211031
View Documents Promotion Details
Cytiva
3030-931
68 cm 58 cm Sheet
690.00CHF
Pack of 100
Only null left
In Stock
Add to basket
11378924
View Documents Promotion Details
Cytiva
3030-866
25 cm 20 cm Sheet
128.00CHF
Pack of 100
Only null left
In Stock
Add to basket
11929197
View Documents
Cytiva
3030-6132
14 cm 12 cm Sheet
33.65CHF
Pack of 100
Only null left
In Stock
Add to basket
11939197
View Documents
Cytiva
3030-6185
14 cm 11 cm Sheet
51.25CHF
Pack of 100
Only null left
In Stock
Add to basket
11328934
View Documents
Cytiva
3030-6187
15.2 cm 20.3 cm Sheet This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 618 41 11 for assistance
N/A
11949197
View Documents Promotion Details
Cytiva
3030-6188
20 cm 15 cm Sheet
71.60CHF
Pack of 100
Only null left
In Stock
Add to basket
10154153
View Documents Promotion Details
Cytiva
3030-6189
10.16 cm 13.34 cm Sheet
32.40CHF
Pack of 100
Only null left
In Stock
Add to basket
11955477
View Documents
Cytiva
3030-6461
41 cm 26 cm Sheet
255.00CHF
Pack of 100
Only null left
In Stock
Add to basket
11948915
View Documents
Cytiva
3030-347
43 cm 35 cm Sheet
310.00CHF
Pack of 100
Only null left
In Stock
Add to basket
Description
Description
Manufactured entirely from high-quality cotton linters with no additives and tested specifically for chromatographic techniques. Whatman™ Grade 3MM Chr Chromatography Paper ensures the wicking capability and uniformity of capillary action that are important in chemical separations.
  • Smooth surface
  • No “paper making” additives—the final organic purity of these chromatography grade papers represents the highest level that can be achieved without oxidation and degradation of the cellulose fibers
  • Flowrate: 130mm/30 min. (water)
  • Medium thickness: 0.34mm
  • Widely used for electrophoresis, general chromatography, and blotting applications
Specifications
Specifications
0.34 mm Whatman medium thickness 3 MM Chr paper
0.34 mm
For Electrophoresis, General Chromatography, and Blotting AppliCatIons
Cellulose
130 mm/30 min.
Smooth
3 MM Chr
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva Whatman™ 3MM Chr Chromatography Paper