Learn More
CKS1B, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CKS1B.
Marke: Abnova PAB26892.100uL
Beschreibung
CKS1B protein binds to the catalytic subunit of the cyclin dependent kinases and is essential for their biological function. The CKS1B mRNA is found to be expressed in different patterns through the cell cycle in HeLa cells, which reflects a specialized role for the encoded protein. At least two transcript variants have been identified for this gene, and it appears that only one of them encodes a protein. [provided by RefSeq
Spezifikation
CKS1B | |
Polyclonal | |
Unconjugated | |
Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
CKS1B | |
CKS1/PNAS-16/PNAS-18/ckshs1 | |
Rabbit | |
Affinity purification | |
RUO | |
1163 | |
Human, Mouse | |
Liquid |
Immunofluorescence, Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of CKS1B. | |
In PBS, pH 7.2 (0.05% sodium azide) | |
P61024 | |
CKS1B | |
A synthetic peptide corresponding to human CKS1B. | |
100 μL | |
Primary | |
CKS1B polyclonal antibody detects endogenous levels of CKS1B protein. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only