Learn More
CDCA8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA590486
455.00 CHF gültig bis 2025-03-16
Benutzen Sie den Promotions-Code "24070" um Ihren Promotions-Preis zu erhalten.
Benachrichtigungen:
Um den Rabatt von 33.33% zu erhalten, müssen Kunden drei Einheiten desselben Produkts zum Listenpreis in einer einzigen Bestellung kaufen. Es gibt keine Begrenzung für die Anzahl von 3er Sets, die Kunden kaufen können. PCODE: 24111
Beschreibung
Immunogen sequence: MAPRKGSSRV AKTNSLRRRK LASFLKDFDR EVEIRIKQIE SDRQNLLKEV DNLYNIEILR LPKALREMNW LDYFALGGNK QALEEAATAD LDITEINKLT AEAIQTPLKS AKTRKVIQVD EMIV; Positive Samples: A-549, 293T, Mouse testis, Rat brain; Cellular Location: Chromosome, Cytoplasm, Nucleus, centromere, cytoskeleton, nucleolus, spindle
CDCA8 is a component of a chromosomal passenger complex (CPC) required for stability of the bipolar mitotic spindle. The chromosomal passenger complex, which includes Survivin, CDCA8, INCENP and Aurora-B, is known to play crucial roles during mitosis and cell division. It was found that CDCA8 interacting with Aurora-B, INCENP and Survivin, increases during G2/M phase and then reduces after exit from M phase. CDCA8 is cell cycle regulated, down-regulated in response to p53/Rb-signaling, and up-regulated in many types of cancerous tissues. In Drosophila cells, inactivation of CDCA8 results in polyploidy, delayed mitosis and abnormal tissue development, indicating its critical role for cell proliferation. Recent studies show that CDCA8 is essential for cell proliferation during early embryonic development, and its early embryonic lethality cannot be rescued by the loss of p53. Its aberrant expression is linked to a poor prognosis for gastric cancer.
Spezifikation
CDCA8 | |
Polyclonal | |
Unconjugated | |
CDCA8 | |
4831429J16Rik, AU044747, BOR, Borealin, Cdca8, cell division cycle associated 8, cell division cycle-associated protein 8, D4Ertd421e, Dasra B, DasraB, dasra-B, embryonic stem cell-related, hDasra-B, MESrg, MESRGP, PESCRG3, Pluripotent embryonic stem cell-related gene 3 protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
500545, 52276, 55143 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1.88 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal, pH 7.3 | |
Q53HL2, Q6AXW0, Q8BHX3 | |
CDCA8 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-124 of human CDCA8 (NP_0605711) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Produktvorschläge
Kunden, die diesen Artikel ansahen, interessierten sich auch für
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.