Learn More
WNK1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 28357-1-AP-150UL
Beschreibung
The WNK1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human WNK1. This antibody recognizes human antigen. The WNK1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spezifikation
WNK1 | |
Polyclonal | |
Unconjugated | |
150 μL | |
65125 | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
367 μg/mL | |
Rabbit | |
Primary | |
Human |