Learn More
Julabo Valegro Ethernet Activation
Activation of the Valegro Ethernet to allow integration into the network.
Marke: Julabo 30004147
Beschreibung
Activating the Ethernet interface makes it possible to integrate Valegro recirculating coolers into your network and thus gain full access to all their functions via a PC.
The Ethernet interface already installed in the Valegro series can be activated at a later date.
Spezifikation
Ethernet Activation | |
Ethernet | |
Valegro |