missing translation for 'onlineSavingsMsg'
Learn More

Metro™ Super Adjustable Super Erecta™ Gitterregal – verchromt

Richten Sie Ihr Lager entsprechend den Anforderungen Ihres Labors ein. Das einzigartige, offene Design des Super Adjustable Super Erecta Chrom-Gitterregals von Metro ermöglicht viele Konfigurationen und freie Luftzirkulation und minimiert Staubansammlungen.

Marke:  Metro™ A2436NC

 Lernen Sie weitere Varianten dieses

Artikelnummer. 11745669

  • 134.00 CHF / 1 Stück
Nur noch null auf Lager
Verfügbar ab: 27-10-2025
Zum Warenkorb hinzufügen

Includes:

Each shelf shipped with one pack of four nonconductive sleeve/wedges.

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Description

Description

  • Each shelf provides approximately 76% open area for increased light penetration and improved visibility of stored items.
  • Can be loaded and unloaded easily from all sides.
  • Hard, durable surface.
  • Abrasion resistant.
  • Ideal for dry, noncorrosive environments.
  • Shelves adust in 1 in. increments with patented shelf-release corners.
  • Shelf releases and wedges offer improved rigidity.
  • Assembles quickly without tools; assembled units are nonconductive.

Order Info

Pfosten separat erhältlich.

Specifications

Specifications

Wire Shelf
Ideal zum Trocknen, nicht korrosive Umgebungen
Vier nicht leitende Muffen/Keile
91.4 cm
61 cm
91,4 x 61 cm (36 x 24 Zoll)
Metro™ Super Erecta™ verstellbares Regalsystem
36 in.
24 in.
Chrom
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Metro™ Super Adjustable Super Erecta™ Gitterregal – verchromt > 24 x 36 in. (61 x 91cm)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.