Learn More
R&D Systems™ Substance P Parameter Assay Kit
Substance P Parameter Assay Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Marke:  R&D Systems™ SKGE007
 Lernen Sie weitere Varianten dieses
Artikelnummer. 18762394
  • 4000.00 CHF / 1 Set
Verfügbar ab: 17-06-2025
Entdecken Sie weitere Sonderangebote
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
The Substance P Parameter Assay Kit quantifies multi-species Substance P in cell culture supernates (50 ul), serum (50 ul), heparin plasma (50 ul), saliva (50 ul), urine (50 ul).
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
39.00 - 2500pg/mL
Biotin
ELISA-Kit
Substance P Levels in Various Samples
Multi-species
1 Kit
Store the unopened product at -20° to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
43.8pg/mL
Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules.
Cell Culture Supernatant, Plasma (Heparin), Speichel, Serum, Urin
6 x 96-well plates
ELISA
Cell Culture Supernates (50uL),Serum (50uL),Heparin Plasma (50uL),Saliva (50uL),Urine (50uL)
neurokinin 1, neurokinin 2, neurokinin A, Neurokinin-1, neuromedin L, neurokinin alpha, neuropeptide K, neuropeptide gamma), neuropeptide gamma, neuropeptide K, NK2, NKA, NKNAneurokinin alpha, NPK, PPT, protachykinin-1, substance K, substance P, TAC1, TAC2neuromedin L, tachykinin 2, tachykinin, precursor 1, tachykinin, precursor 1 (substance K, substance P, neurokinin 1, neurokinin 2
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
One moment while we fetch your results.
Zertifikate
Sonderangebote
Sonderangebote
Berichtigung von Produktinhalten
Name des Produkts
R&D Systems™ Substance P Parameter Assay Kit > Six 96-well plates