Learn More
Bode Chemie™ Sterillium™
Sterillium™ is the classic among the alcohol-based disinfectants for hygienic and surgical hand disinfection. For 50 years, it has proven its worth in practice and undergone comprehensive scientific reviews.
39.70 CHF - 58.75 CHF
Spezifikation
Produkttyp | Hand Disinfectant |
---|---|
Form | Liquid |
Zur Verwendung mit (Anwendung) | Hygienic and Surgical Hand Disinfection |
Verpackung | Flasche |
Beschreibung
- This classic amongst rub-in hand disinfectants - successful for 50 years. Thanks to its reliable antimicrobial activity and its skin friendliness, it is the favourite of millions of users around the world.
- Bactericidal, mycobactericidal, yeasticidal and virucidal against enveloped viruses
- Safe antimicrobial activity for surgical hand disinfection within 90 seconds
- Very good skin tolerability and replenishing
- Increase the skin's moisture levels with regular use
Spezifikation
Hand Disinfectant | |
Liquid | |
Hygienic and Surgical Hand Disinfection | |
Flasche |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Name des Produkts
Bode Chemie™ Sterillium™