Learn More
Bode Chemie™ Sterillium™
Sterillium™ is the classic among the alcohol-based disinfectants for hygienic and surgical hand disinfection. For 50 years, it has proven its worth in practice and undergone comprehensive scientific reviews.
Marke: Bode Chemie™ 100395
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- This classic amongst rub-in hand disinfectants - successful for 50 years. Thanks to its reliable antimicrobial activity and its skin friendliness, it is the favourite of millions of users around the world.
- Bactericidal, mycobactericidal, yeasticidal and virucidal against enveloped viruses
- Safe antimicrobial activity for surgical hand disinfection within 90 seconds
- Very good skin tolerability and replenishing
- Increase the skin's moisture levels with regular use
Spezifikation
Hand Disinfectant | |
Hygienic and Surgical Hand Disinfection | |
1 L |
Liquid | |
Flasche |
Berichtigung von Produktinhalten
Name des Produkts
Bode Chemie™ Sterillium™ > Sterillium; Size: 1 L
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur