Learn More
Thermo Scientific STACKING KIT HERATHERM 180L
Marke:  Thermo Scientific 50126667
STACKING KIT HERATHERM 180L
Artikelnummer. 10019729
  • 494.46 CHF / 1 Stück
Lager vorrätig
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Videos
  • Thermo Scientific - Heratherm heating and drying ovens
    Thermo Scientific Heratherm heating and drying ovens are efficient, safe and easy to use.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • subtitles off, selected
      Thermo Scientific Heratherm heating and drying ovens
    • Thermo Scientific - Heratherm heating and drying ovens
      Thermo Scientific Heratherm heating and drying ovens are efficient, safe and easy to use.
      Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
      1x
      • Chapters
      • descriptions off, selected
      • subtitles off, selected
        Thermo Scientific Heratherm heating and drying ovens
      Name des Produkts
      Thermo Scientific STACKING KIT HERATHERM 180L >