Learn More
RXRA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech 10631-1-AP-150UL
 Lernen Sie weitere Varianten dieses
Artikelnummer. 16807133
  • 385.92 CHF / 150 Mikroliter
Lager vorrätig
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Retinoic Acid (RA; active metabolite of vitamin A) plays a prominent role in regulating the transition of proliferating precursor cells (such as carcinoma cells and neuronal precursors) to postmitotic differentiated cells. The Retinoid X Receptors (RXRs) family (RXR alpha, beta, and gamma), preferentially bind 9-cis -RA and regulate gene transcription by forming heterodimers with a second family of RA receptors. RAs have been suggested to potentially play a therapeutic role in cervical cancer. RAs are known to play key roles in neuronal development and an increasing body of evidence indicates that retinoid signaling may regulate synaptic plasticity and associated learning and memory behaviors.
Spezifikation
Spezifikation
RXRA
Polyclonal
Unconjugated
RXRA
NR2B1, Retinoid X receptor alpha, retinoid X receptor, alpha, RXRA
Rabbit
Antigen Affinity Chromatography
RUO
20181, 25271, 6256
-20°C
Liquid
Western Blot
0.15 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P19793, P28700, Q05343
RXRA
RXRA Fusion Protein Ag0987
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen
Berichtigung von Produktinhalten
Name des Produkts
RXRA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated