missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat LCN2 Protein

A cDNA sequence encoding the LCN2 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP12546-100ug

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15970769

  • 1785.00 CHF / 100 Mikrogramm
Verfügbar ab: 11-03-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Beschreibung

Beschreibung

Spezifikation

Spezifikation

170496
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSQDSTQNL IPAPPLISVP LQPGFWTERF QGRWFVVGLA ANAVQKERQS RFTMYSTIYE LQEDNSYNVT SILVRGQGCR YWIRTFVPSS RPGQFTLGNI HSYPQIQSYD VQVADTDYDQ FAMVFFQKTS ENKQYFKVTL YGRTKGLSDE LKERFVSFAK SLGLKDNNIV FSVPTDQCID N
Greater than 95.0% as determined by SDS-PAGE.
LCN2 Protein
Research Use Only
Lcn2
Rat
His
LCN2 protein solution (0.25 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 20% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Rat LCN2 Protein > 100μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.