missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Protein A/G Protein
Recombinant Protein
Marke: Novus Biologicals™ NBP2-34862
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Protein A/G is a genetically engineered protein containing 5 IgG-binding regions of protein A and 2 of protein G. Cell wall binding region, cell membrane binding region and albumin binding region have been removed from the Protein A/G to ensure the maximum specific IgG binding. Protein A/G is ideal for purification of polyclonal or monoclonal IgG antibodies. It binds to various human, mouse and rat IgG subclasses and to total IgG from cow, goat, sheep, house, rabbit, guinea pig, pig, dog and cat.
Spezifikation
Functional Assay, PAGE | |
47.8 kDa | |
Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. | |
Dissolve in distilled water or saline. | |
>95%, by SDS-PAGE and HPLC |
Lyophilized from additive free solution. | |
1 mg | |
Less than 0.1 EU/μg of Protein A/G as determined by LAL method. | |
Biologically Active Proteins | |
Protein A/G |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Novus Biologicals™ Recombinant Protein A/G Protein > 1 mg
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur