Learn More
R&D Systems™ Recombinant Mouse Erythropoietin R Fc Chimera Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Marke: R&D Systems™ 1390-ER-050
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Recombinant Mouse Erythropoietin R Fc Chimera Protein is derived from NS0. The Recombinant Mouse Erythropoietin R Fc Chimera Protein has been validated for the following applications: Bioactivity.
Spezifikation
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS. | |
51.3 kDa (monomer) | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
13857 | |
Erythropoietin R Fc Chimera | |
Mouse myeloma cell line,NS0-derived mouse Erythropoietin R protein, (N-terminus) Mouse Epo R (Ala25-Pro249) Accession &Num; P14753 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) | |
Disulfide-linked homodimer | |
Recombinant |
Berichtigung von Produktinhalten
Name des Produkts
R&D Systems™ Recombinant Mouse Erythropoietin R Fc Chimera Protein > 50ug; Carrier Free
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur