missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PSMA4 Protein
A cDNA sequence encoding the PSMA4 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 3905.00 CHF
Spezifikation
Name | PSMA4 Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15992759
|
enQuireBio™
QP13183-5ug |
5 μg |
131.00 CHF
5 Mikrogramm |
Verfügbar ab: 29-10-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15982759
|
enQuireBio™
QP13183-20ug |
20 μg |
203.00 CHF
20 Mikrogramm |
Verfügbar ab: 29-10-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15972759
|
enQuireBio™
QP13183-1mg |
1 mg |
3905.00 CHF
1 Milligramm |
Verfügbar ab: 29-10-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
PSMA4 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PSMA4 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 1mM DTT and 0.1mM PMSF. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMSRRYD SRTTIFSPEG RLYQVEYAME AIGHAGTCLG ILANDGVLLA AERRNIHKLL DEVFFSEKIY KLNEDMACSV AGITSDANVL TNELRLIAQR YLLQYQEPIP CEQLVTALCD IKQAYTQFGG KRPFGVSLLY IGWDKHYGFQ LYQSDPSGNY GGWKATCIGN NSAAAVSMLK QDYKEGEMTL KSALALAIKV LNKTMDVSKL SAEKVEIATL TRENGKTVIR VLKQKEVEQL IKKHEEEEAK AEREKKEKEQ KEKDK | |
Greater than 95% as determined by SDS-PAGE. |