missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human DNA Polymerase iota GST (N-Term) Protein
Recombinant Protein
Marke: Novus Biologicals™ H00011201-P01-25ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Spezifikation
Western Blot, ELISA, Protein Array, Immunoaffinity Purification | |
11201 | |
25 μg | |
DNA polymerase iota, EC 2.7.7.7, Eta2, polymerase (DNA directed) iota, RAD30 homolog B, RAD30Bpolymerase (DNA-directed), iota, RAD3OB | |
DNA Polymerases, DNA Repair | |
DNA Polymerase iota |
50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. | |
MolecularWeight-theroretical: 106.7 kDa | |
Store at -80°C. Avoid freeze-thaw cycles. | |
POLI | |
>80% by SDS-PAGE and Coomassie blue staining |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Novus Biologicals™ Recombinant Human DNA Polymerase iota GST (N-Term) Protein > 25 μg
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur