Learn More
PRMT2 Mouse anti-Human, Unconjugated, Clone: 1F1B9, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66885-1-IG-20UL
Beschreibung
The PRMT2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PRMT2. This antibody recognizes human, mouse, rat antigen. The PRMT2 antibody has been validated for the following applications: WB, ELISA analysis.
Spezifikation
PRMT2 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot | |
1F1B9 | |
Unconjugated | |
20 μL | |
3275 | |
IgG2a |