missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Podocalyxin (PODXL) (Hematopoietic Stem Cell Marker) Mouse, Clone: PODXL/2185, enQuireBio™
Mouse Monoclonal Antibody
Spezifikation
Antigen | Podocalyxin (PODXL) (Hematopoietic Stem Cell Marker) |
---|---|
Klon | PODXL/2185 |
Konzentration | 200μg/mL |
Anwendungen | Immunohistochemistry |
Klassifikation | Monoclonal |
Spezifikation
Podocalyxin (PODXL) (Hematopoietic Stem Cell Marker) | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant fragment of human Podocalyxin (apprx. aa 310-447) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
165-170kDa |
PODXL/2185 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
5420 | |
IgG1 | |
Protein A/G purified from Bioreactor concentrate. | |
Cell Surface and Cytoplasmic | |
HeLa, Raji, Jurkat cells, Angiosarcoma, Breast, Prostate, Liver, Pancreatic, Kidney |