missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCD1 / PD1 / CD279 Rabbit, Clone: PDCD1/1410R, enQuireBio™
Rabbit Monoclonal Antibody
234.00 CHF - 484.00 CHF
Spezifikation
Antigen | PDCD1 / PD1 / CD279 |
---|---|
Klon | PDCD1/1410R |
Konzentration | 200μg/mL |
Anwendungen | Immunohistochemistry |
Klassifikation | Monoclonal |
Spezifikation
PDCD1 / PD1 / CD279 | |
200μg/mL | |
Monoclonal | |
Rabbit | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
5133 | |
Human recombinant PDCD1 protein fragment | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
55kDa |
PDCD1/1410R | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_016859782 | |
PDCD1 | |
IgG | |
Protein A/G Purified | |
Cell Surface and Cytoplasmic | |
TY cells or Tonsil. |