missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCD1 / PD1 / CD279 Rabbit, Clone: PDCD1/1410R, enQuireBio™
Rabbit Monoclonal Antibody
Marke: EnquireBio 5133-RBM4-P1
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
PDCD1 / PD1 / CD279 | |
Cell Surface and Cytoplasmic | |
PDCD1/1410R | |
Unconjugated | |
NP_016859782 | |
Rabbit | |
55kDa | |
Protein A/G Purified | |
RUO | |
5133 | |
IgG |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
PDCD1 | |
Human recombinant PDCD1 protein fragment | |
TY cells or Tonsil. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |