missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ornithine Decarboxylase Antibody (SR1904), Novus Biologicals™
Rabbit Monoclonal Antibody
Marke: Novus Biologicals NBP3-22078-50ul
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Ornithine Decarboxylase Monoclonal antibody specifically detects Ornithine Decarboxylase in Human, Mouse, Rat samples. It is validated for Western Blot
Spezifikation
Ornithine Decarboxylase | |
Monoclonal | |
Unconjugated | |
PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
SR1904 | |
Western Blot 1:500-1:2000 | |
ODCEC 4.1.1.17, ornithine decarboxylase, ornithine decarboxylase 1 | |
A synthesized peptide derived from human Ornithine Decarboxylase (Uniprot #: P11926) | |
50 μg | |
Cell Cycle and Replication | |
4953 | |
Store at -20°C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ornithine Decarboxylase Antibody (SR1904), Novus Biologicals™ > 50 μg, Unconjugated
Spot an opportunity for improvement?Share a Content Correction