Learn More
R&D Systems™ Mouse MDL-1/CLEC5A (NP_001033693) VersaClone cDNA
Sequence verified insert equipped with translated sequences for the insert and entire plasmid DNA.
Marke: R&D Systems™ RDC2597
Beschreibung
- The Mouse MDL-1/CLEC5A (NP_001033693) VersaClone cDNA clone contains the complete ORF for MDL-1/CLEC5A, along with a Kozak consensus sequence for optimal translation initiation
- The gene insert is flanked with convenient multiple cloning sites, which can be used to easily cut and transfer the gene cassette into your desired expression vector
- The MDL-1/CLEC5A insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
Spezifikation
mMDL-1/CLEC5A | |
10 μg at 0.2 μg/μL | |
23601 | |
Mouse |
NP_001033693 | |
DNA is provided in 10 mM Tris-Cl, pH 8.5 | |
Store the unopened product at -20 to -80°C | |
10 μg |