Learn More
R&D Systems™ Mouse MDL-1/CLEC5A (NP_001033693) VersaClone cDNA
Sequence verified insert equipped with translated sequences for the insert and entire plasmid DNA.
Marke:  R&D Systems™ RDC2597
Artikelnummer. 18762308
  • 328.00 CHF / 10 Mikrogramm
Verfügbar ab: 11-06-2025
Entdecken Sie weitere Sonderangebote
Beschreibung
  • The Mouse MDL-1/CLEC5A (NP_001033693) VersaClone cDNA clone contains the complete ORF for MDL-1/CLEC5A, along with a Kozak consensus sequence for optimal translation initiation
  • The gene insert is flanked with convenient multiple cloning sites, which can be used to easily cut and transfer the gene cassette into your desired expression vector
  • The MDL-1/CLEC5A insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
mMDL-1/CLEC5A
10 μg at 0.2 μg/μL
23601
Mouse
NP_001033693
DNA is provided in 10 mM Tris-Cl, pH 8.5
Store the unopened product at -20 to -80°C
10 μg
Name des Produkts
R&D Systems™ Mouse MDL-1/CLEC5A (NP_001033693) VersaClone cDNA > Quantity: 10 μg