Learn More
Novus Biologicals™ Met-enkephalin Recombinant Protein Antigen
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Marke:  Novus Biologicals™ NBP1-90944PEP
Artikelnummer. 18778043
  • 291.00 CHF / 0.10 Milliliter
Verfügbar ab: 10-06-2025
Entdecken Sie weitere Sonderangebote
Beschreibung
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human PENK. The Met-enkephalin Recombinant Protein Antigen is derived from E. coli. The Met-enkephalin Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
This is a blocking peptide for NBP1-90944. This is a human recombinant protein expressed in E. coli with a N-terminal His-ABP tag and purified with IMAC chromatography.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
5179
Chromatography
Unmarkiert
ECSQDCATCSYRLVRPADINFLACVMECEGKLPSLKIWETCKELLQLSKPELPQDGTSTLRENSKPEESHLLAKRYGGFMKRYGGFMKKMDELYPMEPEEEANGSEILAKRYGGFMKKDAEE
Human
PENK
32kDa
Name des Produkts
Novus Biologicals™ Met-enkephalin Recombinant Protein Antigen > 0.1mL
Nur für Forschungszwecke