Learn More
Novus Biologicals™ Met-enkephalin Recombinant Protein Antigen
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Marke: Novus Biologicals™ NBP1-90944PEP
Beschreibung
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human PENK. The Met-enkephalin Recombinant Protein Antigen is derived from E. coli. The Met-enkephalin Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.This is a blocking peptide for NBP1-90944. This is a human recombinant protein expressed in E. coli with a N-terminal His-ABP tag and purified with IMAC chromatography.
Spezifikation
5179 | |
Chromatography | |
Unmarkiert | |
ECSQDCATCSYRLVRPADINFLACVMECEGKLPSLKIWETCKELLQLSKPELPQDGTSTLRENSKPEESHLLAKRYGGFMKRYGGFMKKMDELYPMEPEEEANGSEILAKRYGGFMKKDAEE |
Human | |
PENK | |
32kDa |
Nur für Forschungszwecke