Learn More
Brannan™ LO-tox™ Lab Thermometer with 3-points Traceable Calibration
LO-tox™ lab thermometer complete with calibration certificate.
Marke: Brannan™ 44/826/8 + 49/101/0
Beschreibung
- LO-tox™ is an organic filling specially chosen to achieve a wide temperature span and provide the lowest practical levels of hazard
- Safe non-mercury filling
- Supplied with calibration certificate
- Non-roll end cap
- Supplied as standard in protective individual, resealable telescopic tube
- Made in UK
Spezifikation
0°C, 50°C, 90°C | |
Teilweises Eintauchen | |
Glass | |
680°F | |
14 °F | |
14°F to 680°F | |
± 2.0°C at 90°C | |
76 mm |
UK Traceable to National Standards ISO 9001 | |
30.5 cm | |
Brauner Alkohol | |
+360°C | |
-10 °C | |
-10°C to +360°C | |
2 °C | |
1.2 cm |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.