Learn More
LAMR1 RPSA Mouse anti-Human, Unconjugated, Clone: 2A9A1, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 67324-1-IG-150UL
Beschreibung
The LAMR1, RPSA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human LAMR1, RPSA. This antibody recognizes human, mouse, rat antigen. The LAMR1, RPSA antibody has been validated for the following applications: WB, ELISA analysis.
Spezifikation
LAMR1 RPSA | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot | |
2A9A1 | |
Unconjugated | |
150 μL | |
3921 | |
IgM |