Beschreibung
Chemsplash Jet Gown with Taped Seams
- Material: 88GSM Chemsplash Jet Spray Fabric
- High Hazard Protection
- Ankle Length and Tie Back
- Double Cuffs - Internally Knitted and Elasticated Outside
- Double Stand Collar
- Velcro and Adhesive Tape Fastening on Inside Rear Neck
- Anti-Static and Non-Linting
- Taped seams
- PFAS Free
Description: The Chemsplash Jet Gown with Taped Seams, Type PB(3-B), is highly resistant against high hazard permeation, offering high levels of protection against even the most corrosive chemicals. Made from a thick 88gsm heavy weight Polypropylene/Polyethylene multilayer laminated material. Chemsplash accessories are designed to protect the wearer from contaminants in the environment or to protect the environment and production from body-shedding particles or hair from the wearer. This range has been designed to ensure maximum comfort and wearability while maintaining the highest safety standards.
- Umweltvorteile umfassen:
- Recycelter Inhalt
Spezifikation
Spezifikation
Größe | Universal |
Geschlecht | Unisex |
Anzahl Taschen | 1 |
Produkttyp | Controlled Environments Gown |
Material | Composite Fabric |
Farbe | Yellow |
Verschlusstyp | Ties |
Einwegprodukt | Single-use |
Sterilität | Non-sterile |
Stulpenform | Knit |