Learn More
R&D Systems™ ITS Media Supplement (100X)
Supports cell proliferation under low serum conditions. R&D Systems ITS Media Supplement (100X) is a pre-optimized and serum-free media supplement that helps to maintain culture media consistency across passages and experiments.
Marke:  R&D Systems™ AR013
Artikelnummer. 18748237
  • 80.00 CHF / 5 Milliliter
Verfügbar ab: 10-03-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
  • Contains optimized concentrations of insulin, transferrin and selenite
  • Chemically-defined media decreases experimental variation
  • High lot-to-lot consistency decreases variation
  • Performance tested on multiple cell lines
  • QC-tested on multiple cell lines
Precautions
  • Transferrin was tested at the donor level using an FDA licensed method and found to be non-reactive for anti-HIV-1/2 and Hepatitis B surface antigen
  • As no testing can offer complete assurance of freedom from infectious reagents, these reagents should be handled as if capable of transmitting infection
Limitations
  • The safety and efficacy of the product in diagnostic or other clinical uses has not been established
  • The reagent should not be used beyond the expiration date indicated on the label
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
5mL ITS Media Supplement (100X). Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
ITS (Insulin, Transferrin, Selen)
5 mL
RUO
Dokumentation
One moment while we fetch your results.
Name des Produkts
R&D Systems™ ITS Media Supplement (100X) > 5mL
For laboratory research use only. Not for use in diagnostic procedures.