Learn More
Eppendorf epT.I.P.S.™ Motion as Reload System
Eppendorf epT.I.P.S. Motion work with the epMotion platform, with a TipHolder accessory the reload trays offer an environmentally friendly option.
Marke: Eppendorf 0030014545
278.00 CHF gültig bis 2025-12-31
Benutzen Sie den Promotions-Code "25339" um Ihren Promotions-Preis zu erhalten.
Beschreibung
- epT.I.P.S. Motion can be used with epMotion™ 96 or integrated into your workstation as a reload system when combined with the tip holder
- The high-quality reload tips are delivered in a PET thermoformed tray with sealed lid. Upgrading to the autoclavable, aluminum tip holder, which replaces the disposable box, provides waste reduction of up to 40%
- Eppendorf consumables and thus also epT.I.P.S. Motion pipette tips as Reload System are shipped in packaging consisting of at least 62% recycled cardboard
Spezifikation
10 μL | |
Unsteril | |
Nicht autoklavierbar | |
Hanging Tip Rack | |
Roboterspitzen | |
Eppendorf epMotion | |
Polypropylen |
Ungefiltert | |
klar | |
For Eppendorf epMotion series | |
Standard, Straight | |
Single-use | |
No |
Produktvorschläge
Kunden, die diesen Artikel ansahen, interessierten sich auch für
Sonderangebote
-
Mehrere Hersteller
Präzision an Ihren Fingerspitzen
Liquid Handling mühelos meistern.Aktionscode: 24122Gültig bis zum: 2025-04-13
Neues Angebot Sparen Sie bis zu 92%