missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX21 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NB100-1718JF646
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
DDX21 Polyclonal antibody specifically detects DDX21 in Human, Mouse, Zebrafish samples. It is validated for Western Blot, ChIP assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), ChIP assay (ChIP), KnockDown
Spezifikation
DDX21 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
9188 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ChIP Assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Janelia Fluor 646 | |
DEAD (Asp-Glu-Ala-Asp) box polypeptide 21, DEAD box protein 21, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 21, DKFZp686F21172, EC 3.6.1, EC 3.6.4.13, Gu protein, GUA, gu-alpha, GURDB, nucleolar RNA helicase 2, Nucleolar RNA helicase Gu, Nucleolar RNA helicase II, RH II/Gu, RH-II/GU, RH-II/GuA, RNA helicase II/Gu alpha | |
The immunogen recognized by this antibody maps to a region between residue 725 and the C-terminus (residue 783) of human DEAD/H (Asp-Glu-Ala-Asp/His) Box Polypeptide 21 using the numbering given in entry NP_004719.2 (GeneID 9188). | |
0.1 mL | |
Primary | |
Human, Mouse, Zebrafish | |
Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
DDX21 Antibody, Janelia Fluor™ 646, Novus Biologicals™ > 0.1 mL; Janelia Fluor 646
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur