missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DACH1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 10914-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human DACH1.
This gene encodes a member of the diaphanous subfamily of the formin family. Members of this family are involved in actin remodeling and regulate cell movement and adhesion. Mutations in this gene are associated with autosomal domit auditory neuropathy 1. Multiple transcript variants encoding different isoforms have been found for this gene.Spezifikation
DACH1 | |
Polyclonal | |
Unconjugated | |
DACH1 | |
DACH, DACH1, Dachshund homolog 1, Dachshund-1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13134, 1602, 306096 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9QYB2, Q9UI36 | |
DACH1 | |
DACH1 Fusion Protein Ag1354 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
DACH1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur