Learn More
Cytochrome C Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA589231
443.80 CHF gültig bis 2025-02-13
Benutzen Sie den Promotions-Code "24070" um Ihren Promotions-Preis zu erhalten.
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Immunogen sequence: MGDVEKGKKI FIMKCSQCHT VEKGGKHKTG PNLHGLFGRK TGQAPGYSYT AANKNKGIIW GEDTLMEYLE NPKKYIPGTK MIFVGIKKKE ERADLIAYLK KATNE; Positive Samples: SH-SY5Y, 293T, BT-474, 22Rv1, THP-1, Mouse heart, Mouse kidney, Rat kidney; Cellular Location: Mitochondrion intermembrane space
Cytochrome C is an electron transporting protein that resides within the intermembrane space of the mitochondria, where it plays a critical role in oxidative phosphorylation and the production of cellular ATP. Release of cytochrome C from the mitochondria to the cytosol is triggered by apoptotic stimuli. In the cytoplasm, the cytochrome C binds apoptotic protease activating factor which activates the apoptotic initiator procaspase 9. Cytosolic cytochrome C functions in the activation of caspase 3, an ICE family molecule that is a key effector of apoptosis.Spezifikation
Cytochrome C | |
Polyclonal | |
Unconjugated | |
Cycs | |
CG17903, CG17903-PA, CG17903-PB, CRF, CYC, CYC2, Cycs, CYCSA, Cyct-c-p, Cyt c, cyt c-p, cyt.c, CytC, Cyt-c, CYTC2, Cyt-c2, CytC-2, cyt-c-p, cytC-p, Cyt-c-p-PA, Cyt-c-p-PB, Cytochrome c, cytochrome c DC4, Cytochrome c proximal, Cytochrome C, expressed in somatic tissues, cytochrome c, somatic, Cytochrome c-2, cytochrome c-like, cytochrome c-like protein, Cytochrome c-proximal, cytochrome-c-p, Cytochrome-c-proximal, DC4, dCyt.C-p, DMc01, Dmel\CG17903, Dmel_CG17903, hc1, HCS, l(2)k13905, lethal (2) k13905, LOC100341175, LOC100357840, MGC128356 protein, pDMc01, RH17228p, THC4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
13063, 25309, 54205 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.8 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide, pH 7.3 | |
P62897, P62898, P99999 | |
Cycs | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-105 of human Cytochrome C (NP_0618201) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Produktvorschläge
Kunden, die diesen Artikel ansahen, interessierten sich auch für
Viewing 1-5 of 9
Berichtigung von Produktinhalten
Name des Produkts
Cytochrome C Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur