missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Grant Instruments Controlled Rate Freeze Thaw (CRFT)

Controlled Rate Freeze Thaw (CRFT) for cryopreservation. Interchangeable heads must be ordered seperately.
Marke: Grant Instruments CRFT EU
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Controlled rate freezer and thawer that maximizes cell survival without using liquid nitrogen.
Applications
- Transgenic embryology research
- Stem cell research
- Clinical and research samples, e.g. lymphocytes and tissue cell lines in conventional cryovials
- Various mammalian cells including cardiomyocytes, adipose, liver and muscle
- Cord blood derived stem cells
- Adherent cells and stem cells in microplates
- Cell suspensions in numbered/barcoded arrays
- System integrations
- Suitable for applications in veterinarian IVF
Key features and benefits
- Accurate and reproducible control of cooling rates and sample temperatures
- A focus on maximising cell survival rates
- Controlled and customisable freezing and thawing
- Linear and non-linear freezing profiles
- Liquid nitrogen, alcohol and cryogen free - no need for 'topping up'
- Interchangeable heads for flexibility (ordered separately)
- Free of charge software allows you to design, track, export and report temperature profiles
- Easy to use and samples can be nucleated/seeded in-situ
- Low running costs: estimated at 1% of liquid nitrogen-controlled rate freezing and runs on a 24 V supply
- Temperature range of -100°C to 30°C
- Servicing and calibration options available
- Command protocols available

Spezifikation
Cryogenic Freezer/Thawer | |
Plastic | |
Plastic | |
Benchtop, CRFT | |
Weiß | |
2 Jahre | |
Nein | |
290 x 345 x 380 mm | |
Password Protected Software | |
Yes | |
USB and Software Supplied | |
EU | |
50/60 Hz | |
Up to 55 | |
Helium | |
11.7 kg | |
8.9 kg |
1 | |
Aluminium | |
CE | |
No | |
Vials, Straws, PCR Plates | |
Controlled Thawing | |
290 x 345 x 460 mm | |
Internal Data Logger USB | |
Via Software | |
UPS (As Optional Extra) | |
3.5 A | |
100/240 V | |
170 W | |
Air-cooled | |
25.8 lb. | |
19.62 lb. | |
Interchangeable Heads |
Produktvorschläge
Kunden, die diesen Artikel ansahen, interessierten sich auch für
Viewing 1-4 of
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Grant Instruments Controlled Rate Freeze Thaw (CRFT) > EU Plug
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur