missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cIAP1 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1H3F1, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66626-1-IG-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
cIAP1 encodes a protein that is a member of a family of proteins that inhibits apoptosis by binding to tumor necrosis factor receptor-associated factors TRAF1 and TRAF2, probably by interfering with activation of ICE-like proteases. This encoded protein inhibits apoptosis induced by serum deprivation and menadione, a potent inducer of free radicals.Spezifikation
cIAP1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q13490, Q62210 | |
BIRC2 | |
cIAP1 Fusion Protein Ag21398 | |
20 μL | |
Primary | |
Human, Porcine, Mouse, Rat | |
Antibody | |
IgG1 |
Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
1H3F1 | |
Unconjugated | |
BIRC2 | |
API1, BIRC2, C IAP1, cIAP1, Hiap 2, HIAP2, IAP 2, IAP homolog B, IAP2, MIHB, RING finger protein 48, RNF48 | |
Mouse | |
Protein G | |
RUO | |
100622859, 11797, 329, 60371 | |
-20°C | |
Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
cIAP1 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1H3F1, Proteintech > 20 μL; Unconjugated
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur