Learn More
Sampling Systems™ Chemical Spoon and Spatula
This versatile chemical spoon and spatula has a robust design and is made from 304 stainless steel. It can be used as both a spoon and a spatula to allow for greater versatility in the sampling of both powder and liquid products.
Marke:  Sampling Systems™ A625-200
 Lernen Sie weitere Varianten dieses
Artikelnummer. 17962012
  • 6.60 CHF / 1 Stück
Nur noch null auf Lager
Verfügbar ab: 02-07-2025
Beschreibung
  • No sharp edges or crevices, making it ideal for use in food, pharma and chemical companies
  • Available in a range of sizes
  • Heavy duty design for comfortable use
  • Ideal for measuring, sampling and dosing powders and liquids
Spezifikation
Spezifikation
Löffel
Autoklavierbar
No
201 mm
Stainless Steel
Reusable
Unsteril
Zertifikate
Aktionen
Aktionen
Berichtigung von Produktinhalten
Name des Produkts
Sampling Systems™ Chemical Spoon and Spatula > Chemical Spoon and Spatula; Length (Metric): 201 mm