missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 delta Mouse anti-Human, Mouse, Clone: 3A12H2, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 60194-1-IG-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The protein encoded by this gene is part of the T-cell receptor/CD3 complex (TCR/CD3 complex) and is involved in T-cell development and signal transduction. The encoded membrane protein represents the delta subunit of the CD3 complex, and along with four other CD3 subunits, binds either TCR alpha/beta or TCR gamma/delta to form the TCR/CD3 complex on the surface of T-cells. Defects in this gene are a cause of severe combined immunodeficiency autosomal recessive T-cell-negative/B-cell-positive/NK-cell-positive (SCIDBNK). Two transcript variants encoding different isoforms have been found for this gene. Other variants may also exist, but the full-length natures of their transcripts has yet to be defined.Spezifikation
CD3 delta | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P04234, P04235 | |
Cd3d | |
CD3 delta Fusion Protein Ag10207 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Western Blot | |
3A12H2 | |
Unconjugated | |
CD3D | |
CD3 delta, CD3D, CD3-delta, T cell receptor T3 delta chain, T3D | |
Mouse | |
Protein G | |
RUO | |
12500, 915 | |
-20°C | |
Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
CD3 delta Mouse anti-Human, Mouse, Clone: 3A12H2, Proteintech > 150 μL; Unconjugated
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur