Learn More
Brady™ Polyesteretikettenkartuschen für BMP™21-PLUS- und BMP™21-LAB-Drucker
Zur Verwendung mit BMP™21-PLUS- und BMP™21-LAB-Druckern
Marke: Brady™ 139746
Beschreibung
- For smooth surface indoor labeling
- Made of white or clear self laminating polyester material
- Glossy finish and permanent acrylic adhesive
- Compatible with Brady M210 Label Printer, Brady M210-LAB Label Printer, Brady M211 Label Printer, BMP21 LAB Label Printer, BMP21 Label Printer, BMP21-PLUS Label Printer, IDPAL Handheld Label Maker, LABPAL Handheld Label Maker
- M21-250-423, M21-250-430 and M21-250-430-WT-CL are compatible only with the BMP™21-PLUS printer
Applications:
Asset Tracking, Barcoding, Component ID, Equipment ID, General ID, General Identification, General Purpose ID, Location ID, Outlet/Jack ID, Patch Panel, Patch Panel - ID Port, Plate, Rating Plates, DataComm Equipment
Compliance
RoHS-konform, UL-anerkannt, CSA-zugelassen, AGA-zugelassen, halogenfrei (DIN VDE 0472/Teile 815)
Spezifikation
Etiketten-Kassette | |
Brady M210 Etikettendrucker, Brady M210-LAB Etikettendrucker, Brady M211 Etikettendrucker, BMP21-LAB Etikettendrucker, BMP21-PLUS Etikettendrucker | |
Thermotransfer | |
0,25 in. | |
6.35 mm |
Clear | |
Polyester (B-430) | |
21 ft. | |
6.4 m | |
M21-250-430-WT-CL |