missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aurora B Mouse, Clone: AURKB/1521, enQuireBio™
Mouse Monoclonal Antibody
234.00 CHF - 484.00 CHF
Spezifikation
Antigen | Aurora B |
---|---|
Klon | AURKB/1521 |
Konzentration | 200μg/mL |
Anwendungen | Western Blot |
Klassifikation | Monoclonal |
Spezifikation
Aurora B | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
9212 | |
Recombinant human Aurora B protein fragment (aa89-251) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
39kDa |
AURKB/1521 | |
Western Blot | |
Unconjugated | |
RUO | |
NP_001300884 | |
AURKB | |
IgG2b | |
Protein A/G Purified | |
Nuclear and Cytoplasmic | |
HeLa cells. Tonsil, Placenta, Hodgkin's Lymphoma or Colon Carcinoma |